Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ WH-4-023

Click to view available options
Quantity:
10 mg
50 mg
Description
WH-4-023 is a potent and selective Lck and Src inhibitor (IC50 values are 2 and 6 nM respectively). Exhibits >300-fold selectivity against p38α and KDR. Also potently inhibits SIK (IC50 values are 10, 22 and 60 nM for SIK 1, 2 and 3 respectively) and displays selectivity over a range of closely related kinases. Supports self-renewal of naive hESCs in combination with PD 0325901, CHIR 99021 and SB 590885 .
Specifications
Specifications
Quantity | 50 mg |
Product Type | WH-4-023 |
Purity | >98% |
Molecular Weight (g/mol) | 568.67 |
Inhibitors | WH-4-023 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction