Learn More
Description
Suitable for filtering serum, plasma and tissue culture solutions—as well as other protein-containing solutions where minimal adsorptive protein loss is required.
- Polypropylene housing
- Autoclavable
- Female Luer lock on inlet side; male slip Luer on outlet side
4mm dia. Filters
- For samples up to 2mL
- Filter area: 0.2cm2 Hold-up volume (with air purge) <7.5μL
- For samples up to 10mL
- Filter area: 1.3cm2
- Hold-up volume (after air purge) <25μL
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Passes USP Class VI requirements for biosafety. |
| Material | Polysulfone |
| Pore Size | 0.2 μm |
| Sterility | Non-sterile |
| For Use With (Application) | For filtering serum, plasma and tissue culture solutions – as well as other protein-containing solutions where minimal adsorptive protein loss is required. |
| Hold up Volume (After Air Purge) | ≤25 μL |
| Inlet Style | Female Luer Lock |
| Outlet Style | Male Luer Slip |
| Quantity | 100/Pk. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.