Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Accessories for Mini-UniPrep™ Syringeless Filters

Catalog No. 09924492 Shop All Cytiva Products
Change view
Click to view available options
Type:
Multi-sample Compressor Stand
Multi-sample Compressor Tray
Single-sample Compressor
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
09-924-492 Multi-sample Compressor Tray
09-924-491 Multi-sample Compressor Stand
09-924-490 Single-sample Compressor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-924-492 Supplier Cytiva Supplier No. MUPG2MCWT8

Used with Mini-UniPrep syringeless plunger filter units for fast and easy HPLC/UHPLC sample preparation. Whatman™ Compressors for Mini-UniPrep™ Syringeless Filters process one or multiple filter vials simultaneously for

Whatman Mini-UniPrep Syringeless Filters integrate an autosampler vial, filtration membrane, plunger, and cap/septa into one consumable product. Compressors activate the plunger to filter the sample through the membrane without hand stress.

  • Compatible with Mini-UniPrep and/or Mini-UniPrep G2 syringeless filter vials
  • Multi-chamber compressors process multiple vials simultaneously for stream-lined filtration
  • Mini-UniPrep filter vials prepare samples 3x faster than standard syringe filters, and can be loaded directly into an autosampler
  • Options include: 1-, 6- or 8-position hand or handle compressors (additional 8-position trays also available)
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description Vial tray for the G2 Multi-compressor (mfr. no. MUPG2MCPWC8)
For Use With (Equipment) Mini-UniPrep G2 Syringeless Filters
For Use With (Application) Sample filtration/preparation for HPLC/UHPLC
Holds 8 Sample Vials
Housing Material Polypropylene
Includes Tray only
Quantity 1 Ea.
Type Multi-sample Compressor Tray
Length (English) 4.33 in.
Length (Metric) 110 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.