Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Anotop™ 10 Sterile Syringe Filters
GSA_VA

Catalog No. 09924566 Shop All Cytiva Products
Change view
Click to view available options
Pore Size:
0.02 μm
0.2 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
09-924-566 0.2 μm
09-924-565 0.02 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-924-566 Supplier Cytiva Supplier No. 68091122
Add to Cart
Add to Cart

Hold-up volume: <30μL (10mm size), <200μL (25mm)

Similar to Anotop filters, but include binder-free, glass microfiber prefilter with Anotop™ inorganic membrane in a polypropylene housing.

Glass Microfiber Prefilter

  • Increases throughput
  • Maintains membrane performance
  • Reduces need for centrifuging before filtering, sequential filtering, or refiltering
  • Permits use of 0.1μm pore size Anotop Plus filters to remove mycoplasma contaminants from tissue culture
Characteristics
  • Hold-up volume: <30μL (10mm size), <200μL (25mm)
  • Pressure limit, 7kg/cm (100psi)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Material Anapore
Material (Membrane) Alumina-based Anopore
Pore Size 0.2 μm
Sterility Sterile
Filtration Area 0.78 cm2
For Use With (Application) HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water
Hold up Volume (After Air Purge) 100 mL
Housing Material Polypropylene (PP)
Inlet Style Female Luer Lock
Outlet Style Male Luer
Pressure Rating 6.9 bar (100 psi)
Quantity 50/Pk.
Temperature Range (English) 113°F
Temperature Range (Metric) 45°C
Tip Material Polypropylene
Type Syringe Filter
Diameter (Metric) 10 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.