Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Anotop™ 10mm Syringe Filters

Catalog No. 099261 Shop All Cytiva Products
Change view
Click to view available options
Pore Size:
0.02 μm
0.1 μm
0.2 μm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
09-926-1 0.2 μm
09-926-2 0.1 μm
09-926-3 0.02 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-926-1 Supplier Cytiva Supplier No. 68091022
Add to Cart
Add to Cart

Removes virtually 100% of particulates larger than pore size. Whatman™ Anatop™ 10mm Syringe Filters include a highly defined capillary pore structure and narrow pore-size distribution, enabling a high level of consistency in particle retention.

Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes up to 5mL (10mm diameter filter) or 100mL (25mm diameter filter).

  • Nondeformable, honeycombed Anopore™ membrane
  • Inherently hydrophilic membrane is free of adhesives and surfactants
  • Low protein binding, low fluid retention for maximum sample recovery
  • Unidirectional flow

Order Info

Hexagonal, thermal-welded polypropylene housing is free of adhesives and binders.

TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Material (Membrane) Alumina-based Anopore
Pore Size 0.2 μm
Sterility Non-sterile
Filtration Area 0.78 cm2
For Use With (Application) HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water
Hold up Volume (After Air Purge) <20 μL
Housing Material Polypropylene
Inlet Style Female Luer Lock
Outlet Style Male Luer Slip
Pressure Rating 6.9bar (100psi)
Quantity 50/Pk.
Temperature (Metric) Max 45°C
Type Syringe Filter
Volume (Metric) Up to 10 mL
Diameter (Metric) 10 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.