Learn More
Description
Often used for removing particulates from potentially hazardous samples. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial.
- Preassembled units consist of a filter barrel and a plunger
- Built-in air purge minimizes holdup
- Integrated construction eliminates filter pop-off
- Support stand protects slip-Luer tip, recessed to reduce chance of contamination
- Remove particulates from samples up to 5mL in volume
- Filtration area: 1.7cm2
Glass Microfiber (GMF)
- High loading capacity, binder-free, fast flowrates; for clarifying aqueous or organic solutions, pH 3 to 8
- Nylon (NYL)
- Inherently hydrophilic; for clarifying aqueous and organic samples, pH 3 to 10
- Polytetrafluoroethylene (PTFE)
- Hydrophobic; for samples >50% organic solvents
- Polyvinylidene Fluoride (PVDF)
- Low protein binding; for aqueous samples
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Material (Membrane) | PTFE |
| Pore Size | 0.2 μm |
| Sterility | Non-sterile |
| Quantity | 50/Pk. |
| Type | Syringeless Filter |
| Volume (Metric) | ≤5 mL |
| Diameter (Metric) | 13 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
