Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Autovial™ 5mL Syringeless Filters

Catalog No. 0992118 Shop All Cytiva Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Material (Membrane):
Glass Microfiber
Nylon
PTFE
PVDF
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Material (Membrane)
09-921-18 0.45 μm PTFE
09-921-13 0.2 μm PTFE
09-921-15 0.45 μm PVDF
09-921-20 0.45 μm Glass Microfiber
09-921-17 0.45 μm Nylon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 09-921-18 Supplier Cytiva Supplier No. AV115NPUORG
Add to Cart
Add to Cart

Replaces syringe-coupled devices with single filter units for one-time use. Whatman™ Autovial™ 5mL Syringeless Filters are pre-assembled filtration devices for removing particulates from samples.

Often used for removing particulates from potentially hazardous samples. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial.

  • Preassembled units consist of a filter barrel and a plunger
  • Built-in air purge minimizes holdup
  • Integrated construction eliminates filter pop-off
  • Support stand protects slip-Luer tip, recessed to reduce chance of contamination
  • Remove particulates from samples up to 5mL in volume
  • Filtration area: 1.7cm2

Glass Microfiber (GMF)

  • High loading capacity, binder-free, fast flowrates; for clarifying aqueous or organic solutions, pH 3 to 8
  • Nylon (NYL)
  • Inherently hydrophilic; for clarifying aqueous and organic samples, pH 3 to 10
  • Polytetrafluoroethylene (PTFE)
  • Hydrophobic; for samples >50% organic solvents
  • Polyvinylidene Fluoride (PVDF)
  • Low protein binding; for aqueous samples
TRUSTED_SUSTAINABILITY

Specifications

Material (Membrane) PTFE
Pore Size 0.45 μm
Sterility Non-sterile
Quantity 50/Pk.
Type Syringeless Filter
Volume (Metric) ≤5 mL
Diameter (Metric) 13 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.