Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Autovial™ Syringeless Filters: 12mL Capacity, Nonsterile

Catalog No. 09800989 Shop All Cytiva Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Material (Membrane):
Cellulose Acetate
Glass Microfiber
Nylon
PTFE
PVDF
Quantity:
1000/Pk.
50/Pk.
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Material (Membrane) Quantity
09-800-989 0.45 μm Nylon 1000/Pk.
05-713-720 0.45 μm PVDF 1000/Pk.
09-916A 0.45 μm PVDF 50/Pk.
09-916B 0.2 μm PVDF 50/Pk.
09-917A 0.45 μm Nylon 50/Pk.
09-917B 0.2 μm Nylon 50/Pk.
09-918B 0.2 μm PTFE 50/Pk.
09-918A 0.45 μm PTFE 50/Pk.
09-919 0.45 μm Glass Microfiber 50/Pk.
22-042627 0.45 μm PTFE 1000/Pk.
09-921-4C 0.45 μm Cellulose Acetate 50/Pk.
09-921-6A 0.2 μm - 50/Pk.
09-921-6B 0.45 μm - 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 09-800-989 Supplier Cytiva Supplier No. AV525UNAO
Add to Cart
Add to Cart
Request a Quote

Faster sample handling, up to 12mL in volume

Replace syringe-coupled devices with single filter units for one-time use. Often used for removing particulates from potentially hazardous samples. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial.

  • Preassembled units consist of a filter barrel and a plunger
  • Built-in air purge minimizes holdup
  • Integrated construction eliminates filter pop-off
  • Support stand protects slip-Luer tip, recessed to reduce chance of contamination
  • Handle samples up to 12mL in volume
  • All have an integral 25mm diameter filter in barrel (graduated in 1mL intervals) with polyethylene plunger
  • Filtration area: 3.0cm2
  • Glass microfiber prefilter ensures higher flow with less clogging
  • Units with NYL, PTFE, and PVDF membranes are available sterile

Cellulose Acetate (CA)

  • Low nonspecific protein binding, high loading capacity; for biologicals
Glass Microfiber (GMF)
  • High loading capacity, binder-free, fast flowrates; for clarifying aqueous or organic solutions, pH 3 to 8
Nylon (NYL)
  • Inherently hydrophilic; for clarifying aqueous and organic samples, pH 3 to 10
Polypropylene (PP)
  • For aqueous and organic solutions; ideal for aggressive samples and high debris
Polytetrafluoroethylene (PTFE)
  • Hydrophobic; for samples >50% organic solvents
Polyvinylidene Fluoride (PVDF)
  • Low protein binding; for aqueous samples

TRUSTED_SUSTAINABILITY

Specifications

Material (Membrane) Nylon
Pore Size 0.45 μm
Sterility Non-sterile
Prefilter Glass
Quantity 1000/Pk.
Type Syringeless Filter
Volume (Metric) ≤12 mL
Diameter (Metric) 25 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.