Learn More
Description
- Streamlines syringeless filtration by filtering up to eight filter vials simultaneously
- Designed for Cytiva's Mini-UniPrep™ Standard Plastic Filter Vial
- Mini-UniPrep™ Vials can be loaded directly into the autosampler
- The multiprocessor includes one blue 8-position tray
- Additional trays provide the added convenience of pre-loading up to eight Mini-UniPrep™ Filter Vials
Specifications
Specifications
| Color | Blue |
| For Use With (Application) | HPLC/UHPLC |
| Product Line | Whatman |
| Quantity | 1 Ea. |
| Type | Tray |
| Dimensions (L x W x H) | 110 x 110 x 45 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.