Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.2μm BA83 Protran

Catalog No. 09301398 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50/Pk.
Format:
Disc
Sheet
Size (Metric):
140 x 140 mm
82 mm dia.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Size (Metric)
09-301-398 50/Pk. Sheet 140 x 140 mm
09-301-112 50/Pk. Disc 82 mm dia.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-301-398 Supplier Cytiva Supplier No. 10401403
Add to Cart
Add to Cart

100% pure nitrocellulose to ensure the highest binding capacity possible

  • Universal membrane suitable for Western, Southern, and Northern blotting
  • Hydrophilic, so no prewetting with methanol is required
  • High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
  • Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
  • Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
  • Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
  • Also available in circular formats

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 140 mm
Width (Metric) 140 mm
For Use With (Application) Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection
Binding Capacity High
Quantity 50/Pk.
Format Sheet
Material Nitrocellulose
Pore Size 0.2 μm
Size (Metric) 140 x 140 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.