Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.2μm BA83 Protran

Catalog No. p-4909551
Click to view available options
Quantity:
50/Pk.
Format:
Disc
Sheet
Size (Metric):
140 x 140 mm
82 mm dia.
This item is not returnable. View return policy
This item is not returnable. View return policy

100% pure nitrocellulose to ensure the highest binding capacity possible

  • Universal membrane suitable for Western, Southern, and Northern blotting
  • Hydrophilic, so no prewetting with methanol is required
  • High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
  • Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
  • Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
  • Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
  • Also available in circular formats

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection
Binding Capacity High
Quantity 50/Pk.
Format Disc
Material Nitrocellulose
Pore Size 0.2 μm
Size (Metric) 82 mm dia.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.