Learn More
Cytiva Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Circles
Shop All Cytiva Products

Description
- Universal membrane suitable for Western, Southern, and Northern blotting
- Hydrophilic, so no prewetting with methanol is required
- High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
- Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
- Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
- Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| For Use With (Application) | Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection |
| Binding Capacity | High |
| Diameter (Metric) | 82 mm |
| Quantity | 50/Pk. |
| Format | Disc |
| Material | Nitrocellulose |
| Pore Size | 0.45 μm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.