Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Circles

Catalog No. 09301129 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100/Pk.
1000/Pk.
50/Pk.
Diameter (Metric):
25 mm
82 mm
132 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Diameter (Metric)
09-301-129 100/Pk. 25 mm
09-301-102 50/Pk. 82 mm
09-301-130 50/Pk. 132 mm
09-301-132 1000/Pk. 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 09-301-129 Supplier Cytiva Supplier No. 10402506
Add to Cart
Add to Cart

100% pure nitrocellulose to ensure the highest binding capacity possible

  • Universal membrane suitable for Western, Southern, and Northern blotting
  • Hydrophilic, so no prewetting with methanol is required
  • High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
  • Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
  • Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
  • Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection
Binding Capacity High
Diameter (Metric) 25 mm
Quantity 100/Pk.
Format Disc
Material Nitrocellulose
Pore Size 0.45 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.