Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Qualitative Filter Paper: Grade 1 Circles

Catalog No. 09805J Shop All Cytiva Products
Click to view available options
Diameter (Metric):
10 mm
15 mm
20 mm
25 mm
30 mm
32 mm
42.5 mm
47 mm
55 mm
70 mm
85 mm
90 mm
110 mm
125 mm
150 mm
185 mm
240 mm
270 mm
320 mm
385 mm
400 mm
Quantity:
100/Pk.
400/Pk.
500/Pk.

Catalog No. 09-805J

Supplier: Cytiva 1001240

Only null left
Add to Cart
Add to Cart

Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Circles are widely used standard grade filter papers for routine applications with medium retention and flow rates.

 

These cellulose filters are used in qualitative analytical techniques to determine and identify materials and for clarifying liquids. 
  • Particle Retention: >11μm
  • Porosity: medium
  • Filtration speed: ASTM™, 40 sec.; Herzberg, 150 sec.
  • Surface: smooth
  • Separates precipitates such as lead sulfate, calcium oxalate (hot) and calcium carbonate
  • Used for soil analysis and seed testing procedures in the agricultural industry
  • Used to separate solid foodstuffs from associated liquid or extract liquid in the food industry
  • Atmospheric dust is collected from airflow and the stain-intensity measured 
  • Impregnated with a chromogenic reagent and color formation quantified by optical reflectance for gas detection

 


TRUSTED_SUSTAINABILITY

Specifications

Airflow Rate 13sec./100mL/sq. in.
Basis Weight 87 g/m2
Description Grade 1 Qualitative Filter Paper Standard Grade, circle, 240 mm
Format Disc
Material Cellulose
Particle Retention >11 μm
Filtration Speed 150 sec.
Flow Rate 57mL/min. (water)
For Use With (Application) A widely used standard grade filter paper for routine applications with medium retention and flow rate. It covers a wide range of laboratory applications and is frequently used for clarifying liquids.
Grade Grade 1
Porosity Medium
Quantity 100/Pk.
Thickness (Metric) 180 μm
Type Qualitative Filter Paper
Diameter (English) 9.44 in.
Diameter (Metric) 240 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.