Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Qualitative Filter Paper: Grade 1 Circles
Shop All Cytiva Products
Click to view available options
Diameter (Metric):
10 mm
15 mm
20 mm
25 mm
30 mm
32 mm
42.5 mm
47 mm
55 mm
70 mm
85 mm
90 mm
110 mm
125 mm
150 mm
185 mm
240 mm
270 mm
320 mm
385 mm
400 mm
Quantity:
100/Pk.
400/Pk.
500/Pk.
Description
Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Circles are widely used standard grade filter papers for routine applications with medium retention and flow rates.
These cellulose filters are used in qualitative analytical techniques to determine and identify materials and for clarifying liquids.
- Particle Retention: >11μm
- Porosity: medium
- Filtration speed: ASTM™, 40 sec.; Herzberg, 150 sec.
- Surface: smooth
- Separates precipitates such as lead sulfate, calcium oxalate (hot) and calcium carbonate
- Used for soil analysis and seed testing procedures in the agricultural industry
- Used to separate solid foodstuffs from associated liquid or extract liquid in the food industry
- Atmospheric dust is collected from airflow and the stain-intensity measured
- Impregnated with a chromogenic reagent and color formation quantified by optical reflectance for gas detection

Specifications
Specifications
Airflow Rate | 13sec./100mL/sq. in. |
Basis Weight | 87 g/m2 |
Description | Grade 1 Qualitative Filter Paper Standard Grade, circle, 15 mm |
Format | Disc |
Material | Cellulose |
Particle Retention | >11 μm |
Filtration Speed | 150 sec. |
Flow Rate | 57mL/min. (water) |
For Use With (Application) | A widely used standard grade filter paper for routine applications with medium retention and flow rate. It covers a wide range of laboratory applications and is frequently used for clarifying liquids. |
Grade | Grade 1 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction