Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ UniFilter™ Microplates, 24-Well, 10mL

Catalog No. p-155888 Shop All Cytiva Products
Click to view available options
Pore Size:
1.2 μm
10 to 12 μm
25 to 30 μm
Color:
Clear
Natural

Ideal for applications that require larger sample or reagent volumes (e.g. biomolecular purification by solid-phase extraction and organic synthesis

  • Natural polypropylene plates
  • Patented integral filter design prevents well-to-well cross contamination
  • Standard footprint—compatible with most plate readers, autosamplers and robots
  • Unique drip director design on the bottom of the plates controls filtrate placement
  • Available in several media types

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Description The 10 mL UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation.
Array 4 x 6
Pore Size 25 to 30 μm
Color Clear
Product Type UNIFILTER Microplate, 24-well, 10 mL, 25 to 30 μm melt blown polypropylene, natural polypropylene
Material Polypropylene
Volume (Metric) Well 10,000 μL
No. of Wells 24
Sterility Non-sterile
Well Shape Filter
For Use With (Application) Biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation
Includes Long drip director and MBPP Filter
Quantity 25/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.