Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences 520 Bottle-Top Dispenser Reservoirs
SDP

Catalog No. p-47637 Shop All DWK Life Sciences Products
Click to view available options
Volume (Metric):
250 mL
500 mL
1000 mL
2500 mL
Color:
Amber
Clear
Diameter (Metric) Outer Neck:
32 mm
45 mm
Shape:
Round
Square

Large selection of glass and Polyethylene reagent bottles, each supplied with one screw cap. Perfectly adapted with Calibrex™ bottle top dispensers.

  • Corresponding neck adapters supplied with the dispensers.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Reagent Bottle
Color Amber
Material Soda Lime Glass
Volume (Metric) 1000 mL
For Use With (Equipment) 520 model bottletop dispenser
Shape Square
Volume (English) 33.81 oz.
Diameter (English) Outer Neck 1.77 in.
Diameter (Metric) Outer Neck 45 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.