Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences 520 Bottle-Top Dispenser Reservoirs
Shop All DWK Life Sciences Products

Click to view available options
Volume (Metric):
250 mL
500 mL
1000 mL
2500 mL
Color:
Amber
Clear
Diameter (Metric) Outer Neck:
32 mm
45 mm
Shape:
Round
Square
Description
- Corresponding neck adapters supplied with the dispensers.

Specifications
Specifications
Product Type | Reagent Bottle |
Color | Amber |
Material | Soda Lime Glass |
Volume (Metric) | 2500 mL |
For Use With (Equipment) | 520 model bottletop dispenser |
Shape | Round |
Volume (English) | 84.53 oz. |
Diameter (English) Outer Neck | 1.77 in. |
Diameter (Metric) Outer Neck | 45 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction