Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ M-T Vial File™ and Accessories
GSA_VASDP

Catalog No. p-3207814 Shop All DWK Life Sciences Products
Click to view available options
Product Type:
Alphanumerical Index Card
Index Card
M-T Vial File
Partition
Storage Box
Quantity:
35 Case
6 Case
For Use With (Equipment):
20mL M-T vial file
228778 Hinged Case
228780 Hinged Case
24 x 20mL vials
40 x 4mL vials
54 x 8mL vials
60 x 2mL vials
W228790 Hinged Case
W228792 Hinged Case

For compact and organized sample storage

  • Stores vials in clear polystyrene case
  • Foam inserts and partitions provide protection
  • With alphanumerical indexing card for identification
  • Index cards and partitions also available separately

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Polystyrene
For Use With (Application) Vial storage and transport
For Use With (Equipment) 60 x 2mL vials
Includes Clear polystyrene hinged case, partitions, foam inserts and alphanumerical indexing card
No. of Places 60
Product Type M-T Vial File
Quantity 6 Case
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.