Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Maxwell's Wheel

Shop All United Scientific Supplies, Inc Products


Click to view available options
Grade:
5 to 12
Description
This classic device is used to demonstrate the conservation of mechanical energy and its conversion from one form to another.
- After the wheel is rotated by hand to roll the supporting cords to the top and then released, its potential energy turns into kinetic energy of rotation as it falls
- When the wheel reaches its bottom position, the kinetic energy changes back to potential energy as it rolls the cords up again
- This process continues until all the energy is lost to friction
- All metal construction
- Includes activity guide
Specifications
Specifications
| Grade | 5 to 12 |
| Format | Kit |
| Languages | English |
| Subject | Physics |
Warning: Cancer and reproductive harm
Prop 65
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction