Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WIC1
GREENER_CHOICE

Catalog No. p-7250160
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 75-801-0

Supplier: Tocris Bioscience™ 7580/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibitor of Wnt/β-catenin signaling

WIC1 is an inhibitor of the canonical Wnt/β-catenin signaling pathway. In vitro, it reduces ABSC (Airway Basal Stem Cell) proliferation and induces ciliated epithelial cell differentiation, in a model of squamous lung cancer. WIC1 also represses the phosphorylation of β-cateninY489 and Tp63 expression.

Specifications

Chemical Name or Material N-[4-(4-Ethyl-1-piperazinyl)phenyl]-2-oxo-2 H-1-benzopyran-3-carboxamide
CAS 943083-58-7
Quantity 10 mg
Target beta-Catenin
Molecular Formula C22H23N3O3
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.