Learn More
Thermco Products Wifi Vaccine Temperature Logger
Description
- This CDC and VFC compliant, ISO/IEC 17025 NIST-traceable 30 day temperature data logger with a display for monitoring vaccine storage in refrigerators or freezers.
- Automatically uploads real-time temperature data to a LogTag Online cloud account via WiFi.
- Non-volatile memory stores up to 16,129 temperature readings.
- Statistical memory tracks maximum and minimum temperatures and alarm durations for each of the last 30 days.
- Users can view and reset minimum and maximum readings at any time.
- Audible and flashing LED alerts provide immediate notification of temperature excursions.
- Data can be downloaded using LogTag Analyzer software or generated as a PDF without software if WiFi is unavailable.
- External buffer bottle probe measures temperatures from -40°C to +99°C (-40°F to +210°F) for accurate vaccine monitoring.
Specifications
Specifications
| Type | Temperature Datalogger |
| Display Type | LCD |
| Alarm Types | Audible, LED Indicator, SMS, Voice |
| Model | Temperature Logger A |