Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ WIKI4

Click to view available options
Quantity:
5 mg
25 mg
Description
WIKI4 is an inhibitor of Wnt/β-catenin signaling (EC50 ∼ 75 nM). Inhibits auto-ADP-ribosylation of tankyrase 2 (TNKS2) (IC50 ∼15 nM); prevents the ubiquitination and degradation of axin. WIKI4 inhibits expression of β-catenin target genes in cancer cell lines as well as in human embryonic stem cells (ESC).
WIKI4 synthesized to Ancillary Material Grade also available.
Specifications
Specifications
Chemical Name or Material | 2-[3-[[4-(4-Methoxyphenyl)-5-(4-pyridinyl)-4H-1,2,4-triazol-3-yl]thio]propyl]-1H-benz[de]isoquinoline-1,3(2H)-dione |
CAS | 838818-26-1 |
Assay Percent Range | >98% |
Quantity | 5 mg |
Molecular Formula | C29H23N5O3S |
Formula Weight | Observed MW: 521.59 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction