Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Wilm's Tumor Protein (WT1) Rabbit Polyclonal Antibody, with BSA and azide

Epredia™ Wilm's Tumor Protein (WT1), Rabbit Polyclonal Antibody provides accurate, reproducible results.

Specifications

Antigen Wilms Tumor Protein (WT1)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
RB9267R1
View Documents
Epredia™
RB9267R1
Prediluted 1 mL N/A N/A N/A
Description

Description

Wilm's Tumor (WT), a sporadic and familial childhood kidney tumor, is genetically heterogeneous. Wilm's tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed infetal kidney and mesothelioma, and its expression has been suggested as a marker for Wilm's tumor and mesothelioma.

Host Species: Rabbit

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of human Wilm's tumor protein

Molecular Weight: 47-55kDa

Positive Control: Wilm's tumor

Cellular Localization: Cytoplasmic and nuclear This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Wilms Tumor Protein (WT1)
Polyclonal
Rabbit
Cancer and Tumor Biology
A synthetic peptide derived from the C-terminus of human Wilms tumor protein
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.