Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ Disposable NMR Tube Caps, 1.7mm Dia., Red

Catalog No. 16800211 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Click to view available options
Color:
Red
When using Chloroform or Acetone as a reference solution, please avoid direct contact between the sample and the disposable cap, and avoid long term storage over 2 hours. If these conditions cannot be avoided, please use PTFE tube caps.
When using Chloroform or Acetone as a reference solution, please avoid direct contact between the sample and the disposable cap, and avoid long term storage over 2 hours. If these conditions cannot be avoided, please use PTFE tube caps.

Different colors help track samples based on various reference solvents.

  • Made from Ethylene Vinyl Acetate (EVA) or Polyethylene (PE), disposable caps have different colors to help track samples
  • Choose cap size based on Tube O.D. size

Specifications

Includes 25 per pack
Diameter (Metric) Outer 1.7 mm
Description Model: Wilmad™ 521-R
Material EVA
Color Red
For Use With (Application) NMR Spectroscopy
Disposable Single-use
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.