Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WIPI1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017359 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017359 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017359 Supplier Novus Biologicals Supplier No. NBP282048
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WIPI1 Polyclonal specifically detects WIPI1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen WIPI1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL
Formulation PBS
Gene Alias Atg18, Atg18 protein homolog, ATG18A, FLJ10055, WD repeat domain phosphoinositide-interacting protein 1, WD repeat domain, phosphoinositide interacting 1, WD40 repeat protein interacting with phosphoinositides of 49 kDa, WD40 repeat protein Interacting with phosphoInositides of 49kDa, WIPI 49 kDa, WIPI-1, WIPI-1 alpha, WIPI49ATG18
Gene Symbols WIPI1
Host Species Rabbit
Immunogen WIPI1 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human WIPI1. The immunogen is located within amino acids 350 - 400 of WIPI1. Amino Acid Squence: LGSGTTEENKENDLRPS
Molecular Weight of Antigen 49 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55062
Test Specificity At least two isoforms of WIPI1 are known to exist; this antibody will detect both isoforms.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.