Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WIPI1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB184156 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184156 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184156 Supplier Novus Biologicals Supplier No. NBP282048H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WIPI1 Polyclonal antibody specifically detects WIPI1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen WIPI1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias Atg18, Atg18 protein homolog, ATG18A, FLJ10055, WD repeat domain phosphoinositide-interacting protein 1, WD repeat domain, phosphoinositide interacting 1, WD40 repeat protein interacting with phosphoinositides of 49 kDa, WD40 repeat protein Interacting with phosphoInositides of 49kDa, WIPI 49 kDa, WIPI-1, WIPI-1 alpha, WIPI49ATG18
Host Species Rabbit
Immunogen WIPI1 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human WIPI1. The immunogen is located within amino acids 350 - 400 of WIPI1. Amino Acid Squence: LGSGTTEENKENDLRPS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cell Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55062
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.