Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WIPI2 Antibody (2A2), FITC, Novus Biologicals™
SDP

Catalog No. NB385979 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385979 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385979 Supplier Novus Biologicals Supplier No. NBP308559F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WIPI2 Monoclonal antibody specifically detects WIPI2 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen WIPI2
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 2A2
Conjugate FITC
Formulation PBS
Gene Alias ATG18B, Atg21, CGI-50, DKFZP434J154, DKFZp686P02188, FLJ12979, FLJ14217, FLJ42984, WD repeat domain phosphoinositide-interacting protein 2, WD repeat domain, phosphoinositide interacting 2, WD40 repeat protein interacting with phosphoinositides 2, WIPI-2, WIPI49-like protein 2
Host Species Mouse
Immunogen Recombinant fragment (around aa 418-436) of human WIPI2 protein (exact sequence is proprietary) (Uniprot: Q9Y4P8)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 26100
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.