Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WIPI2 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB184392 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184392 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184392 Supplier Novus Biologicals Supplier No. NBP282059UV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WIPI2 Polyclonal antibody specifically detects WIPI2 in Human, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen WIPI2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias ATG18B, Atg21, CGI-50, DKFZP434J154, DKFZp686P02188, FLJ12979, FLJ14217, FLJ42984, WD repeat domain phosphoinositide-interacting protein 2, WD repeat domain, phosphoinositide interacting 2, WD40 repeat protein interacting with phosphoinositides 2, WIPI-2, WIPI49-like protein 2
Host Species Rabbit
Immunogen Rabbit polyclonal WIPI2 antibody was raised against a 17 amino acid peptide near the amino terminus of human WIPI2. The immunogen is located within the first 50 amino acids of WIPI2. Amino Acid Squence: CANFNQDNTEVKGASRAA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 26100
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.