Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WIZ Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176515 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176515 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176515 Supplier Novus Biologicals Supplier No. NBP176515
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WIZ Polyclonal specifically detects WIZ in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen WIZ
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 2.5 ug/mL, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 2.5 ug/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. AAI44333
Gene Alias widely interspaced zinc finger motifs, widely-interspaced zinc finger motifs, Widely-interspaced zinc finger-containing protein, WIZ zinc finger, Zinc finger protein 803, ZNF803protein Wiz
Gene Symbols WIZ
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human TFF3. The immunogen is located within the last 50 amino acids of WIZ. Amino Acid Squence: NFSKADPPPEESQAPQA
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58525
Test Specificity At least four isoforms of WIZ are known to exist; this antibody will detect all four.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.