Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ PCR Preps DNA Purification System

Catalog No. PRA7170 Shop All Promega Corporation Products
Change view
Click to view available options
:
50 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PR-A7170 50 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-A7170 Supplier Promega Supplier No. A7170
Only null left
Add to Cart
Add to Cart

Includes

Wizard SV Purification Resin, Direct Purification Buffer, Wizard Minicolumns, 3cc syringe barrels

A 15-minute column purification method for purifying PCR-amplified DNA away from contaminants, including primer-dimers and amplification primers. Can also be used to purify DNA fragments from agarose gels.

  • Purifies PCR Products Directly from Reactions
  • 15-minute protocol
  • Purify PCR DNA from reactions or gel slices

Specifications

Content And Storage 15°C to 30°C
For Use With (Application) Simple, reliable way to purify double-stranded PCR-amplified
Includes 50mL Wizard™ PCR preps DNA purification Resin, 5mL direct purification Buffer, 50 Wizard™ minicolumns, 50 3cc syringe barrels, protocol
No. of Reactions 50 Preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.