Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WM 1119
GREENER_CHOICE

Catalog No. p-7161694
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity KAT6A (MOZ) competitive inhibitor

WM 1119 is a high affinity and competitive lysine acetyltransferase 6A (KAT6A/MOZ) inhibitor (Kd = 2 nM), which displays >250-fold affinity for KAT6A compared to KAT5 and KAT7. WM 1119 induces cell-cycle exit and cellular senescence, and inhibits growth of B-cell lymphoma cells in vitro (IC50 = 0.25 μM). WM 1119 also reduces H3K9ac in tumor cells, and decreases tumor burden and spleen weight in a mouse lymphoma model.

Specifications

Chemical Name or Material 2-Fluoro-N'-(3-fluoro-5-(pyridin-2-yl)benzoyl)benzenesulfonohydrazide
CAS 2055397-28-7
Quantity 10 mg
Molecular Formula C18H13F2N3O3S
Formula Weight 389.38
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.