Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNK1 Antibody (26G7), Novus Biologicals™
SDP

Catalog No. NB298475 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298475 100 μL
NB298474 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298475 Supplier Novus Biologicals Supplier No. NBP326251100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

WNK1 Monoclonal antibody specifically detects WNK1 in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen WNK1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 26G7
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EC 2.7.11, EC 2.7.11.1, Erythrocyte 65 kDa protein, HSAN2hereditary sensory neuropathy, type II, HSN2MGC163339, hWNK1, KDPMGC163341, KIAA0344, Kinase deficient protein, p65, PHA2C, PRKWNK1PSK, prostate-derived sterile 20-like kinase, Protein kinase lysine-deficient 1, Protein kinase with no lysine 1, protein kinase, lysine deficient 1, serine/threonine-protein kinase WNK1, WNK lysine deficient protein kinase 1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human WNK1 [UniProt Q9H4A3]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 65125
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.