Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNK1 Antibody (M42-P3B10), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP250364AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250364AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250364AF Supplier Novus Biologicals Supplier No. NBP250364AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WNK1 Monoclonal specifically detects WNK1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen WNK1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone M42-P3B10
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.1, Erythrocyte 65 kDa protein, HSAN2hereditary sensory neuropathy, type II, HSN2MGC163339, hWNK1, KDPMGC163341, KIAA0344, Kinase deficient protein, p65, PHA2C, PRKWNK1PSK, prostate-derived sterile 20-like kinase, Protein kinase lysine-deficient 1, Protein kinase with no lysine 1, protein kinase, lysine deficient 1, serine/threonine-protein kinase WNK1, WNK lysine deficient protein kinase 1
Gene Symbols WNK1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide KSISNPPGSN-peptide immunogen is identical in mouse (Mus musculus), rat (Rattus norvegicus), sheep (Ovis aries), bovine (Bos taurus), canine (Canis familiaris), porcine (Sus scrofa), equine (Equus caballus) and chichen (Gallus gallus).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 65125
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.