Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNK4 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB600284Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600284Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600284Y Supplier Novus Biologicals Supplier No. NB600284AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WNK4 Polyclonal specifically detects WNK4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, ICC/IF (Negative).

Specifications

Antigen WNK4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, ICC/IF (Negative)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q96J92
Gene Alias EC 2.7.11, EC 2.7.11.1, PHA2B, PRKWNK4protein kinase, lysine deficient 4, Protein kinase lysine-deficient 4, Protein kinase with no lysine 4, serine/threonine-protein kinase WNK4, WNK lysine deficient protein kinase 4
Gene Symbols WNK4
Host Species Rabbit
Immunogen A synthetic peptide made to a region of the human WNK4 protein between residues 200-300. [UniProt# Q96J92]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Potassium Channels, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 65266
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.