Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-1 Antibody (13F9), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB120021 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120021 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120021 Supplier Novus Biologicals Supplier No. NBP142739DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Wnt-1 Monoclonal specifically detects Wnt-1 in Human, Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Wnt-1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 13F9
Conjugate DyLight 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias INT1Proto-oncogene Int-1 homolog, proto-oncogene Wnt-1, wingless-type MMTV integration site family, member 1, wingless-type MMTV integration site family, member 1 (oncogene INT1)
Host Species Mouse
Immunogen Synthetic peptide corresponding to an internal region of human Wnt-1 protein. (Uniprot: P04628)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cellular Markers, Oncogenes, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7471
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.