Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB353429 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB353429 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB353429 Supplier Novus Biologicals Supplier No. NBP306123APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Wnt-2 Polyclonal antibody specifically detects Wnt-2 in Human samples. It is validated for Western Blot

Specifications

Antigen Wnt-2
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias INT1L1secreted growth factor, Int-1-like protein 1, Int-1-related protein, IRPprotein Wnt-2, wingless-type MMTV integration site family member 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Wnt-2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7472
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.