Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-2b Rabbit anti-Human, Mouse, Rat, Clone: 9M6O7, Novus Biologicals™
SDP

Catalog No. NB166639 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166639 100 μg
NB166640 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166639 Supplier Novus Biologicals Supplier No. NBP315772100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Wnt-2b Monoclonal antibody specifically detects Wnt-2b in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Wnt-2b
Applications Western Blot
Classification Monoclonal
Clone 9M6O7
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Protein Wnt-13, protein Wnt-2b, wingless-type MMTV integration site family, member 2B, WNT13XWNT2, Xenopus, homolog of, XWNT2wingless-type MMTV integration site family, member 13
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-300 of human Wnt-2b (Q93097).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7482
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.