Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-3a Antibody (217804.2R), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB305078 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305078 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305078 Supplier Novus Biologicals Supplier No. FAB13242T

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

Wnt-3a Monoclonal antibody specifically detects Wnt-3a in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen Wnt-3a
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 217804.2R
Conjugate Alexa Fluor 594
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias MGC119418, MGC119419, MGC119420, protein Wnt-3a, wingless-type MMTV integration site family, member 3A
Host Species Rat
Immunogen E.coli-derived recombinant mouse Wnt-3a, Ser36-Gln75, Trp219-Arg269, Accession # P27467
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 89780
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.