Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-5a Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NBP224752F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224752F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224752F Supplier Novus Biologicals Supplier No. NBP224752F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Wnt-5a Polyclonal specifically detects Wnt-5a in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Wnt-5a
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS with 0.05% Sodium Azide
Gene Alias hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, WNT-5A protein
Gene Symbols WNT5A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 190-230 of human Wnt-5a was used as the immunogen for this antibody, GenBank no NP_003383.2.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer Stem Cells, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7474
Target Species Human, Mouse, Rat, Bovine, Monkey
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.