Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-8b Antibody (HL1549) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294861 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294861 Supplier Novus Biologicals Supplier No. NBP325756
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Wnt-8b Monoclonal antibody specifically detects Wnt-8b in Human samples. It is validated for Western Blot

Specifications

Antigen Wnt-8b
Applications Western Blot
Classification Monoclonal
Clone HL1549
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000
Formulation PBS
Gene Alias protein Wnt-8b, wingless-type MMTV integration site family, member 8B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Wnt-8b. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7479
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.