Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-9b Antibody, Novus Biologicals™
SDP

Catalog No. NBP15793720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15793720 20 μL
NBP157937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15793720 Supplier Novus Biologicals Supplier No. NBP15793720UL

Rabbit Polyclonal Antibody

Wnt-9b Polyclonal specifically detects Wnt-9b in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Wnt-9b
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. O14905
Gene Alias Protein Wnt-14b, wingless-type MMTV integration site family, member 15, wingless-type MMTV integration site family, member 9B, WNT14Bprotein Wnt-9b, WNT15Protein Wnt-15
Gene Symbols WNT9B
Host Species Rabbit
Immunogen Synthetic peptides corresponding to WNT9B (wingless-type MMTV integration site family, member 9B) The peptide sequence was selected from the C terminal of WNT9B. Peptide sequence FRETGQVLKLRYDSAVKVSSATNEALGRLELWAPARQGSLTKGLAPRSGD.
Molecular Weight of Antigen 39 kDa
Purification Method Protein A purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7484
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.