Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-9b Antibody, Novus Biologicals™
SDP

Catalog No. NBP151961 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151961 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP151961 Supplier Novus Biologicals Supplier No. NBP151961
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Wnt-9b Polyclonal specifically detects Wnt-9b in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen Wnt-9b
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_003387.1
Gene Alias Protein Wnt-14b, wingless-type MMTV integration site family, member 15, wingless-type MMTV integration site family, member 9B, WNT14Bprotein Wnt-9b, WNT15Protein Wnt-15
Gene Symbols WNT9B
Host Species Goat
Immunogen Peptide with sequence C-KRGNKDLRARADA corresponding to internal region according to NP_003387.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7484
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse and Rat.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.