Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WNT2 Mouse anti-Human, Mouse, Rat, Clone: 3H7B10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | WNT2 |
---|---|
Clone | 3H7B10 |
Concentration | 1 mg/mL |
Applications | Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7074
![]() ![]() |
Proteintech Group Inc
666561IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
WNT2 is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.Specifications
WNT2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Int 1 like protein 1, Int 1 related protein, INT1L1, IRP, Protein Wnt 2, WNT2 | |
WNT2 | |
IgG1 | |
Protein G | |
Antibody |
3H7B10 | |
Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P09544, P21552 | |
114487, 22413, 7472 | |
WNT2 Fusion Protein Ag25185 | |
Primary | |
-20°C | |
WNT2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title