Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ Women's Khaki Flame Resistant Flex Knit Button Down Shirt

Catalog No. 17103747 Shop All Red Kap Industries Products
Click to view available options
Size:
X-Large
2X-Large

Catalog No. 17-103-747

Supplier: Bulwark™ STG5KH RG XL

Only null left
Add to Cart
Add to Cart

Women's Khaki Flame Resistant Flex Knit Button Down Shirt combines the crisp, professional look of an office work shirt with the comfort of your favorite T-shirt.

  • Lightweight stretch fabric and side seam gussets flex with your body for added comfort and range of movement
  • Two chest pockets with button flaps allows convenient storage
  • Sewn in pencil stall and banded collar
  • Easy-to-spot CAT 2/2112 label on the left sleeve leave no question that you're protected on the job site
  • Hazard : Arc Flash, Flash Fire
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 6.75 oz. (230 g/m2) Flame Resistant Jersey
  • Blend: Body: 93% Cotton/7% Spandex Elastane
  • Protection: ATPV Arc Rating: 8.3
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size X-Large
Gender Women's
Material Cotton, Spandex
Color Khaki
Neck Style Collar
Sleeve Style Long Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 2
Hazard Risk Category Category 2
Arc Flash Rating 8.3
Certifications/Compliance NFPA 70E, NFPA 2112, ASTM F1506
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.