Learn More
Bulwark™ Women's Khaki Flame Resistant Flex Knit Long Sleeve T-Shirt
Shop All Red Kap Industries ProductsDescription
- Bulwark FR Women's Flex Knit Tee gives you the stretch of your favorite T-shirt while keeping you cool and dry.
- The lightweight fabric flexes for added comfort and range of movement, curved flatlock seams for optimal fit, and a dropped back hem for a flattering silhouette.
- CAT 2/2112 left sleeve label leaves no question you're protected on the job site.
- Certifications Level 1: NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Number of Pockets: 1
- Primary Closure: Pull On
- Wash Care: Home Wash
- Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
- Blend: Body: 93% Cotton / 7% Spandex Elastane, Contrast Panels: 93% Cotton / 7% Spandex Elastane
- Pocket: Single chest pocket
- Features: Contrasting pattern on sleeve for athletic look, Lightweight stretch fabric flexes with your body for added comfort and range of movement, Dropped back hem presents a flattering silhouette
- Protection: ATPV Arc Rating: 8.3
- Country of Origin: Imported, Made in USA Fabric
- Gender: Female
- Brand: Bulwark™
Specifications
Specifications
| Size | 3X-Large |
| Gender | Women's |
| Material | Cotton, Spandex |
| Color | Khaki |
| Neck Style | Hemmed Crew |
| Sleeve Style | Long Sleeve |
| Construction | Knit |
| Closure Type | Pull On |
| Disposable | Reusable |
| No. of Pockets | 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.