Learn More
United Scientific Supplies Women's Laboratory Coats
SDPSCI_ED
Catalog No. S139589 Shop All United Scientific Supplies, Inc Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Allows for safety and comfort in the laboratory.
  • Knee-length lab coat made of easy-care, white cotton blend fabric (65% polyester, 35% cotton)
  • One upper left breast pocket, two roomy lower pockets, side pocket slit openings, and a sewn-down back belt
  • Tapered and cut for a better fit than unisex or men's lab coats
Specifications
Product Type General Purpose Lab Coat
Size 2X-Large
Gender Women's
Material Polyester, Cotton
Color White
Closure Type Buttons
Cuff Style Hemmed
Disposable Reusable
Sterility Non-sterile
Resistance Light Liquid Splash
Style Standard Length
No. of Pockets 2
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.