Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wortmannin, >99% purity, For TLC, HPLC analysis, 97.96%, MP Biomedicals™

Catalog No. p-4405852 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 mg
5 mg

Chemical Identifiers

CAS 19545-26-7
Molecular Formula C23H24O8
Molecular Weight (g/mol) 428.44
MDL Number MFCD00133927
InChI Key QDLHCMPXEPAAMD-QAIWCSMKSA-N
Synonym wortmannin, wartmannin, antibiotic sl-2052, unii-xva4o219qw, xva4o219qw, pi 3-kinase inhibitor, 1alpha,11alpha-11-acetyloxy-1-methoxymethyl-2-oxaandrosta-5,8-dieno 6,5,4-bc furan-3,7,17-trione, 1r,3r,5s,9r,18s-18-methoxymethyl-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo 10.6.1.0^ 2,10 .0^ 5,9 .0^ 15,19 nonadeca-2 10 ,12 19 ,14-trien-3-yl acetate, 1s,6br,9as,11r,11br-9a,11b-dimethyl-1-methyloxy methyl-3,6,9-trioxo-1,6,6b,7,8,9,9a,10,11,11b-decahydro-3h-furo 4,3,2-de indeno 4,5-h isochromen-11-yl acetate, wortmanin
PubChem CID 312145
ChEBI CHEBI:52289
IUPAC Name (1R,3R,5S,9R,18S)-18-(methoxymethyl)-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo[10.6.1.0²,¹⁰.0⁵,⁹.0¹⁵,¹⁹]nonadeca-2(10),12(19),14-trien-3-yl acetate
SMILES COC[C@H]1OC(=O)C2=COC3=C2[C@@]1(C)C1=C([C@@H]2CCC(=O)[C@@]2(C)C[C@H]1OC(C)=O)C3=O

Specifications

Color White
Assay Percent Range 0.9796
Quantity 1 mg
Formula Weight 428.4
Percent Purity >99%
Physical Form Powder
Chemical Name or Material Wortmannin, For TLC and HPLC analysis
Recommended Storage Store at -20°C, desiccate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.