Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Woulff Gas Wash Bottles

High quality borosilicate 3.3 glass bottles with two ground joint parallel necks
Supplier: Eisco™ CH0511B
This item is not returnable.
View return policy
Description
Warranty and Services
1 year limited manufacturer warranty
Specifications
250 mL | |
Parallel Necks | |
3.8 cm | |
3.8 cm | |
1 Ea. | |
Clear |
Borosilicate Glass | |
Non-sterile | |
10.2 cm | |
Gas wash | |
Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction