Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WQ 1
GREENER_CHOICE

Catalog No. p-7237098
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective σ1 receptor antagonist; active in vivo

WQ 1 is a high affinity and selective σ1 receptor antagonist (pKi values are 10.28-10.85, 5.81-6.53 and 5.80 at σ1, σ2 and DAT). Displays σ1/σ2 selectivity ratio of 29512. Selectively blocks binge eating without a general inhibition of food intake in rats.

Specifications

Chemical Name or Material 2,3-Dihydro-2-[1-(phenylmethyl)-4-piperidinyl]-4 H-1-benzothiopyran-4-one
CAS 1204401-49-9
Quantity 10 mg
Target Sigma1 Receptor Antagonists
Molecular Formula C21H23NOS
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.